Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethylphenylsilane 98%, ACROS Organics™

Click to view available options
Quantity:
10mL
100mL
500mL
Packaging:
Glass bottle
149-74-6
C7H8Cl2Si
191.13
MFCD00000490
GNEPOXWQWFSSOU-UHFFFAOYSA-N
methylphenyldichlorosilane,dichloro methyl phenylsilane,phenylmethyldichlorosilane,silane, dichloromethylphenyl,dichlor-fenyl-methylsilane,dichloro methyl phenyl silane,dichloromethylphenyl silane,unii-9jr2a9v0ji,9jr2a9v0ji,benzene, dichloromethylsilyl
9006
dichloro-methyl-phenylsilane
C[Si](C1=CC=CC=C1)(Cl)Cl
149-74-6
C7H8Cl2Si
191.13
MFCD00000490
GNEPOXWQWFSSOU-UHFFFAOYSA-N
methylphenyldichlorosilane,dichloro methyl phenylsilane,phenylmethyldichlorosilane,silane, dichloromethylphenyl,dichlor-fenyl-methylsilane,dichloro methyl phenyl silane,dichloromethylphenyl silane,unii-9jr2a9v0ji,9jr2a9v0ji,benzene, dichloromethylsilyl
9006
dichloro-methyl-phenylsilane
C[Si](C1=CC=CC=C1)(Cl)Cl
Boiling Point 205°C
Packaging Glass bottle
Specific Gravity 1.176
Flash Point 82°C
Refractive Index 1.5180 to 1.52
Quantity 100mL
Density 1.1760g/mL
Infrared Spectrum Authentic
Color Colorless
Assay Percent Range 98%
Linear Formula C6H5Si(CH3)Cl2
Solubility Information Solubility in water: reacts with water
Formula Weight 191.13
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Dichloromethylphenylsilane, 98%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 205-746-2

RTECSNumber : VV3530000

TSCA : TSCA