Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dichloromethylphosphine, 90%, Tech., Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.3000g/mL |
| Assay Percent Range | 90% |
| Packaging | Glass bottle |
| Linear Formula | CH3PCl2 |
| Quantity | 10 g |
| Formula Weight | 116.92 |
| Grade | Technical |
| Chemical Name or Material | Dichloromethylphosphine |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Harmful if inhaled. Causes severe skin burns and eye damage. Catches fire spontaneously if exposed to air. Reacts violently with water. Contact with water liberates toxic gas. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not allow contact with air. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction