Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicloxacillin sodium salt monohydrate

Catalog No. AAJ6158103
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAJ6158103

Supplier: Thermo Scientific Chemicals J6158103

Only null left
Add to Cart

CAS: 13412-64-1 | C19H21Cl2N3NaO6S | 513.342 g/mol

Dicloxacillin sodium salt is used in the treatment of staphylococci resistant to penicillin G. Dicloxacillin binds to specific penicillin-binding proteins in the bacterial cell wall and therefore inhibits the last stage of bacterial cell wall synthesis. Cell lysis, mediated by bacterial cell wall autolytic enzymes, is the result. Dicloxacillin may interfere with autolysin inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13412-64-1
Molecular Formula C19H21Cl2N3NaO6S
Molecular Weight (g/mol) 513.342
MDL Number MFCD00056865
InChI Key ASMFXKXNAWVTAY-MBGFYOAHSA-N
PubChem CID 131673926
IUPAC Name (2S,5R,6R)-6-[[3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium;hydrate
SMILES [HH].CC1=C(C(=NO1)C2=C(C=CC=C2Cl)Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.O.[Na]

Specifications

Melting Point 222°C to 225°C
Color White
Quantity 1 g
Merck Index 14,3084
Solubility Information Soluble in water
Formula Weight 492.33
Physical Form Powder
Chemical Name or Material Dicloxacillin sodium salt
Hazard Category H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
EINECSNumber 603-794-2
RTECSNumber XH8925000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.