Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicyclopentadiene diepoxide, 98%, Thermo Scientific Chemicals

Catalog No. p-107431
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-21-0
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00077209
InChI Key BQQUFAMSJAKLNB-UHFFFAOYSA-N
Synonym dicyclopentadiene dioxide, dicyclopentadiene diepoxide, unox epoxide 207, epoxide 207, bicyclopentadiene dioxide, unox 207x, unox 207, dicyclopentadiene dioxide van, 1,2:5,6-diepoxyhexahydro-4,7-methanoindan, 4,7-methanoindan, 1,2:5,6-diepoxyhexahydro
PubChem CID 6673
SMILES C1C2C3CC4C(C3C1C5C2O5)O4

Specifications

Melting Point 185.0°C to 189.0°C
Color Beige-Yellow to White
Boiling Point 326.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: 1.4g/L (20°C)
Formula Weight 164.2
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Dicyclopentadiene diepoxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove
EINECSNumber 201-334-1
RTECSNumber PB9625200
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.