Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dicyclopentenyl acrylate
Click to view available options
Quantity:
25 g
100 g
Description
Dicyclopentenyl acrylate is used in organic synthesis of chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -40°C |
| Color | Brown |
| Density | 1.07 g/mL |
| Flash Point | 131°C (268°F) |
| Quantity | 25 g |
| Solubility Information | Fully miscible in water. |
| Formula Weight | 204.27 |
| Physical Form | Liquid |
| Chemical Name or Material | Dicyclopentenyl acrylate |
Safety and Handling
| missing translation for 'healthHazard1' | H315-H317-H319-H334-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H334-H335-H315-H319-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. |
| missing translation for 'healthHazard3' | P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c |
| EINECSNumber | 251-678-1 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction