Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethanolamine solution, 85% (Laboratory), Fisher Chemical

Catalog No. d45500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
D45-500 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D45-500

Chemical Identifiers

CAS 111-42-2 , 7732-18-5
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
MDL Number MFCD00002843
InChI Key ZBCBWPMODOFKDW-UHFFFAOYSA-N
Synonym diethanolamine, 2,2'-iminodiethanol, diolamine, iminodiethanol, bis 2-hydroxyethyl amine, diethylolamine, n,n-diethanolamine, diethanolamin, 2,2'-dihydroxydiethylamine, ethanol, 2,2'-iminobis
PubChem CID 8113
ChEBI CHEBI:28123
IUPAC Name 2-[(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCNCCO

Specifications

Melting Point 28°C
Color Colorless
Density 1.09g/cm³
Boiling Point 268°C
Flash Point 137°C
Packaging Glass Bottle
Linear Formula (HOCH2CH2)2NH
Quantity 500 mL
UN Number 3267
Vapor Density 3.65g/mol
Formula Weight 105.14
Grade Laboratory
Concentration or Composition (by Analyte or Components) 85% (Diethanolamine solution: 85%, Water: 15%)
Physical Form Liquid
Chemical Name or Material Diethanolamine
Show More Show Less
DOTInformation Class 8: Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.