Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethoxymethane, 99%, stab. with 100ppm BHT

Catalog No. p-7026863
Click to view available options
Quantity:
100 mL
500 mL

CAS: 462-95-3 | C5H12O2 | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-95-3
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00009246
InChI Key KLKFAASOGCDTDT-UHFFFAOYSA-N
Synonym diethoxymethane, ethylal, formaldehyde diethyl acetal, diethylformal, diethoxy methane, methane, diethoxy, ethoxymethoxy ethane, ethoxymethyl ethyl ether, 1,1-diethoxymethane, 3,5-dioxaheptene
PubChem CID 10024
IUPAC Name ethoxymethoxyethane
SMILES CCOCOCC

Specifications

Melting Point °C to 66°C
Density 0.831
Boiling Point 87°C to 88°C
Flash Point -5°C (23°F)
Assay Percent Range 0.99
Refractive Index 1.374
Quantity 100 mL
UN Number UN2373
Beilstein 1697253
Formula Weight 104.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethoxymethane, stab. with 100ppm BHT
Show More Show Less
Hazard Category H225-H311-H315-H319-H335
Hazard Statement GHS H Statement
H225-H311-H315-H319-H335
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIETHOXYMETHANE
EINECSNumber 207-330-6
RTECSNumber PA8500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.