Learn More
Diethyl ether, ACS, 98% min, stab. with 0.001% BHT and 3% ethanol
Description
Widely used as a solvent. Dissolves P, S, Br2 and I2. It is a widely used as a solvent in organic synthesis, and in the preparation of Grignard reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -116°C |
| Color | Colorless |
| Density | 0.713 g/mL |
| Boiling Point | 34.6°C |
| Quantity | 6 x 500 g |
| Flash Point | -45°C (-49°F) |
| Odor | Sweet |
| Physical Form | Liquid |
| Refractive Index | 1.3525 |
| Assay Percent Range | 98% min |
| Show More |
Safety and Handling
| Hazard Category | H224-H302-H305-H335-H336-H373 |
| Hazard Statement | GHS H Statement H224-H302-H336 Extremely flammable liquid and vapor. Harmful if swallowed. May cause drowsiness or dizziness. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P314-P330-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: DIETHYL ETHER |
| EINECSNumber | 200-467-2 |
| RTECSNumber | KI5775000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.