Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene glycol monoethyl ether acetate, 97%, Thermo Scientific™

Catalog No. AA44170K2
Click to view available options
Quantity:
50 mL
1 L

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-15-2
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00041928
InChI Key FPZWZCWUIYYYBU-UHFFFAOYSA-N
Synonym carbitol acetate, 2-2-ethoxyethoxy ethyl acetate, diethylene glycol monoethyl ether acetate, ektasolve de acetate, karbitolacetat, ethoxydiglycol acetate, glycol ether de acetate, diglycol monoethyl ether acetate, ethanol, 2-2-ethoxyethoxy-, acetate, ethyldiglycol acetate
PubChem CID 8165
IUPAC Name 2-(2-ethoxyethoxy)ethyl acetate
SMILES CCOCCOCCOC(C)=O

Specifications

Assay Percent Range 97%
Quantity 1 L
Formula Weight 176.21
Physical Form Liquid
Chemical Name or Material Diethylene glycol monoethyl ether acetate
Hazard Category H319: Causes serious eye irritation.
EINECSNumber 203-940-1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.