Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Chromogenic Dehydrated Media: m-EI Agar Base
Click to view available options
Quantity:
100 g
500 g
Certifications/Compliance:
ISO/DIS, 11014
US, EPA
Description
- Any colonies present will exhibit blue halo within 24 hours of the test
- Conforms to EPA Method 1600
Order Info
Available in dehydrated form.

Specifications
Specifications
Certifications/Compliance | US, EPA |
Color | Light Beige |
Content And Storage | 5°C to 25°C |
Form | Solid |
Manufacturing Origin | United States |
Media Type | m EI Agar |
Formulation | Peptone 10g/L, Sodium Chloride 15g/L, Esculin 1g/L, Cycloheximide 0.05g/L, Sodium Azide 0.15g/L, Yeast extract 30g/L, Indoxyl-beta-D-glucoside 0.75g/L, Agar 15g/L |
For Use With (Application) | The chromogenic detection and enumeration of Enterococci in Water by the single-step membrane filtration technique |
Packaging Type | Bottle |
Quantity | 100 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction