Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Fluid Lactose Medium

Catalog No. p-4869075
Click to view available options
Quantity:
10,000 g
2000 g
500 g
Packaging Type:
Bottle
Pail

For cultivation of Salmonella and coliform organisms in water, foods, dairy and pharmaceutical products

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, CCAM, COMPF, SMD
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Lactose Broth
Product Type Microbiology Media
Formulation Beef extract 3g/L, Peptone 5g/L, Lactose 5g/L
For Use With (Application) Cultivation of Salmonella and Coliform organisms in Water, foods, dairy and pharmaceutical products
Packaging Type Pail
Quantity 10,000 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.