Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: MacConkey Agar

Catalog No. DF0075171 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
Certifications/Compliance:
CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, SMWW, USP
ISO 9001:2008, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, SMWW, USP
This item is not returnable. View return policy
This item is not returnable. View return policy

For differential isolation of enteric bacilli based on lactose fermentation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 9001:2008, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, SMWW, USP
Content And Storage Refrigerate
Form Dehydrated
Media Type MacConkey Agar
Product Type Agar
Formulation Pancreatic digest of Gelatin 17g/L, Peptones (meat and Casein) 3g/L, Lactose 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Neutral red 0.03g/L, Crystal Violet 1mg/L
For Use With (Application) For differential isolation of enteric Bacilli based on Lactose fermentation
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.