Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Nutrient Agar
Shop All BD Diagnostic Systems Products
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                100 g
                            
                        
                            
                                2 kg
                            
                        
                    
                            
                                Packaging Type:
                            
                        
                        
                            
                                Bottle
                            
                        
                            
                                Pail
                            
                        
                    
                            
                                Media Type:
                            
                        
                        
                            
                                Nutrient Agar
                            
                        
                    Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, CCAM, COMPF, ISO, SMWW, USDA | 
| Content And Storage | <30°C | 
| Form | Dehydrated | 
| Media Type | Nutrient Agar | 
| Product Type | Agar | 
| Formulation | Beef extract 3g/L, Peptone 5g/L, Agar 15g/L | 
| For Use With (Application) | General purpose medium for the cultivation of a wide variety of microorganisms | 
| Packaging Type | Pail | 
| Quantity | 2 kg | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction