Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: TT (Tetrathionate) Broth Base, Hajna

Catalog No. p-4869136 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
500 g
2000 g
Packaging Type:
Bottle
Pail
Media Type:
TT Broth Base, Hajna
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective enrichment for Salmonella from food and dairy products

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USDA
Content And Storage Refrigerate
Form Dehydrated
Media Type TT Broth Base, Hajna
Product Type Microbiology Media
Formulation Yeast extract 2g/L, Tryptose 18g/L, Dextrose 0.5g/L, D-Mannitol 2.5g/L, Sodium Desoxycholate 0.5g/L, Sodium Chloride 5g/L, Sodium Thiosulfate 38g/L, Calcium Carbonate 25g/L, Brilliant green 0.01g/L
For Use With (Application) For enriching Salmonella from food and dairy products prior to isolation procedures
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.