Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Sabouraud Dextrose Agar

Catalog No. DF0109082 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging Type:
Bottle
Pail
This item is not returnable. View return policy
This item is not returnable. View return policy

Rendered more selective for fungi by the addition of antimicrobics. BD™ Difco™ Sabouraud Dextrose Agar is a general-purpose medium used in the cultivation of yeasts, molds and aciduric microorganisms. It is also recommended for the testing of cosmetics and food.

Difco™ Sabouraud Dextrose Agar is a dehydrated culture media used in qualitative procedures for cultivation of pathogenic and nonpathogenic fungi, particularly dermatophytes.

  • The addition of antimicrobics is a modification designed to increase bacterial inhibition.
  • The low pH of approximately 5.6 is favorable for the growth of fungi and slightly inhibitory to contaminating bacteria in clinical specimens.
  • General Chapters 61 and 62 of the USP describe test methods for using Sabouraud Dextrose Agar when performing the microbial enumeration tests and tests for isolating Candida albicans from nonsterile pharmaceutical products.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP
Color Beige
Content And Storage 2 to 25°C
Form Dehydrated
Media Type Sabouraud Dextrose Agar
Product Type Agar
Formulation Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 5g/L, Dextrose 40g/L, Agar 15g/L
For Use With (Application) Cultivation of pathogenic and nonpathogenic fungi, particularly dermatophytes
Packaging Type Pail
Quantity 10 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.