Learn More
Description
Includes
Kit: 2 x 125mL anode buffer; 4 x 125mL cathode buffer; 12 tubes agarose sealing solution
- DIGE Gel run with DIGE Buffer Kit results in spot maps with reproducibility compared to homecast polyacrylamide gels, thereby reducing technical variation between gels
- Ready-to-use
- Comes with long shelf-life
- Eliminates the need to make buffers, cast gels and wash glass plates
- Offers quality and performance of the Laemmli system with reproducibility, convenience and time savings
- Should be used with the DIGE Buffer kit
- 12.5% precast low fluorscent polyacrylamide gel
- Cast in low fluorescent glass cassette developed for second dimension 2-D electrophoresis
- Solid
- Dimension: 10 x 7.7 x 0.03 in. (255 x 196 x 1mm)
- Designed two-buffer system for running up to twelve DIGE Gels simultaneously
- Contains the amount of reagents necessary for a single run of up to 12 DIGE Gels
- Liquid
Specifications
Specifications
| Description | DIGE Gel is a precast polyacrylamide gel in a low fluorescent glass cassette. |
| Buffer | Buffer based on piperidinopropionamide (PPA) |
| Color | Colorless |
| Separation Range | 12 to 120 kDa |
| Form | Solid |
| For Use With (Application) | Industrial applications: analytical chemistry, research For differential protein analysis |
| Kit Contents | Running Buffers and sealing solution for attaching immobiline™ DryStrip gels (ipg strips) to the top of the Polyacrylamide gel |
| Quantity | 1 Kit: 2 x 125 mL anode buffer; 4 x 125 mL cathode buffer; 12 tubes agarose sealing solution |
| Storage Requirements | 2-8°C |
| Product Type | DIGE Gel and Buffer Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.