Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diglycyl-Lysine Antibody (GX41), Novus Biologicals™
SDP

Catalog No. NB036582 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036582 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036582 Supplier Novus Biologicals Supplier No. NBP288991
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Diglycyl-Lysine Monoclonal specifically detects Diglycyl-Lysine in Non-species specific samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Diglycyl-Lysine
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone GX41
Conjugate Unconjugated
Formulation PBS
Gene Alias Diglycyl Lysine
Host Species Mouse
Immunogen Diglycine-modified histone from calf thymus, Type III-S, lysine rich fraction.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.