Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digoxigenin Antibody (611621), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB374485 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374485 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374485 Supplier Novus Biologicals Supplier No. FAB7520X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Digoxigenin Monoclonal specifically detects Digoxigenin in Multi-species samples. It is validated for Immunohistochemistry.

Specifications

Antigen Digoxigenin
Applications Immunohistochemistry
Classification Monoclonal
Clone 611621
Conjugate Alexa Fluor 532
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias DIG
Host Species Mouse
Immunogen KLH-coupled Digoxigenin
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Multi-species
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.