Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digoxigenin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7067954
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7520RSP 25 μg
MAB7520R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB7520RSP Supplier R&D Systems Supplier No. MAB7520RSP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Digoxigenin Monoclonal specifically detects Digoxigenin in Multi-Species samples. It is validated for Immunohistochemistry.

Specifications

Antigen Digoxigenin
Applications Immunohistochemistry
Classification Monoclonal
Clone 611621R
Conjugate Unconjugated
Dilution Immunohistochemistry 5-25 ug/mL
Formulation Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias DIG
Host Species Mouse
Immunogen KLH-coupled Digoxigenin
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects Digoxigenin labeled proteins, nucleic acids and Digoxigenin conjugated primary and secondary antibodies.
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.