Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Digoxigenin NHS ester
Click to view available options
Quantity:
1 mg
5 mg
Description
Digoxigenin NHS ester is an NHS ester hapten for labeling amino groups within biomolecules (oligonucleotides or proteins). Digoxigenin NHS ester is also used for in situ hybridization.
Specifications
Specifications
Chemical Name or Material | (3β,5β,12β)-3-[2-[[6-[(2,5-Dioxo-1-pyrrolidinyl)oxy]-6-oxohexyl]amino]-2-oxoethoxy]-12,14-dihydroxycard-20(22)-enolide |
CAS | 129273-26-3 |
Quantity | 5 mg |
Target | General Laboratory Research Reagent |
Molecular Formula | C35H50N2O10 |
Purity | 80% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction