Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Dihydrexidine hydrochloride
GREENER_CHOICE

Catalog No. 088425
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective D1-like agonist

A potent, full efficacy dopamine D1 agonist which shows no agonist activity at peripheral D2 receptors or adrenoceptors at doses which cause maximal stimulation of D1 sites. The compound appears to be fully bioavailable in brain and exhibits profound antiparkinsonism effects in vivo.

Chemical Identifiers

CAS 158704-02-0
Molecular Formula C17H18ClNO2

Specifications

Quantity 25 mg
Formula Weight 303.79
Purity >99%
Molecular Weight (g/mol) 303.786
Chemical Name or Material Dihydrexidine hydrochloride
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.