Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydrofolate Reductase/DHFR Antibody (OTI6G7), FITC, Novus Biologicals™
SDP

Catalog No. NBP270569F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270569F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270569F Supplier Novus Biologicals Supplier No. NBP270569F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dihydrofolate Reductase/DHFR Monoclonal specifically detects Dihydrofolate Reductase/DHFR in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Dihydrofolate Reductase/DHFR
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI6G7
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias DHFRP1, dihydrofolate reductase, DYR, EC 1.5.1.3
Gene Symbols DHFR
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human Dihydrofolate Reductase/DHFR expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1719.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.