Learn More
Dihydrogen hexachloroiridate(IV) hydrate, 99% (metals basis), Ir 38-42%
Description
Dihydrogen hexachloroiridate(IV) hydrate is involved in the electrochemical synthesis of polyaniline on the surface of non-noble metal electrodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Black |
| Physical Form | Lump |
| Quantity | 1 g |
| UN Number | UN3260 |
| Solubility Information | Soluble in alcohol and water |
| IUPAC Name | tetrachloroiridium;hydrate;dihydrochloride |
| Formula Weight | 406.93 (Anhydrous) |
| Percent Purity | 99% |
| Concentration or Composition (by Analyte or Components) | Ir 38 to 42% |
| Assay | (metals basis) |
| Show More |
Safety and Handling
| Hazard Category | H290-H302-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H290-H302 Causes severe skin burns and eye damage. Causes serious eye damage. May be corrosive to metals. Harmful if swallowed. |
| Precautionary Statement | P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
| EINECSNumber | 241-012-8 |
| RTECSNumber | NO3592500 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.