Learn More
Dihydrogen hexahydroxyplatinate(IV), 99.9% (metals basis), Pt 61.0% min
Description
Dihydrogen hexahydroxyplatinate(IV) is used as a precursor to produce platinum dioxide (II), a catalyst of hydrogenation and hydrogenolysis of carbonyl group or C-Cl linkages in acyl chlorides. It also acts as a precursor to produce other platinum compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | (decomposition) |
| Color | Yellow |
| Quantity | 5 g |
| Sensitivity | Hygroscopic |
| Solubility Information | Insoluble in water. |
| Formula Weight | 299.17 |
| Percent Purity | 99.9% |
| Concentration or Composition (by Analyte or Components) | Pt ≤61.0% |
| Physical Form | Crystalline |
| Assay | (metals basis) |
| Show More |
Safety and Handling
| Hazard Category | H315-H317-H319-H335 |
| Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
| Precautionary Statement | P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c |
| EINECSNumber | 257-471-2 |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.