Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Dihydrokainic acid
GREENER_CHOICE

Catalog No. p-4783413
Click to view available options
Quantity:
1 mg
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

EAAT2 (GLT-1)-selective non-transportable inhibitor of L-glutamate and L-aspartate uptake

Dihydrokainic acid is an EAAT2(GLT1)-selective non-transportable inhibitor of L-glutamate and L-aspartate uptake (Ki = 23 μM). 130-fold selective over EAAT1 and EAAT3 (Ki > 3 mM).

Chemical Identifiers

CAS 52497-36-6
Molecular Formula C10H17NO4

Specifications

Quantity 1 mg
Formula Weight 215.25
Purity >98%
Molecular Weight (g/mol) 215.25
Chemical Name or Material Dihydrokainic acid
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.