Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydrolipoamide Dehydrogenase/DLD Antibody (OTI6D5), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP27074A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27074A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27074A32 Supplier Novus Biologicals Supplier No. NBP270574AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dihydrolipoamide Dehydrogenase/DLD Monoclonal specifically detects Dihydrolipoamide Dehydrogenase/DLD in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Dihydrolipoamide Dehydrogenase/DLD
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI6D5
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex), branched chain keto acid dehydrogenase complex, dihydrolipoamide dehydrogenase (E3 component of pyruvate dehydrogenase complex, dihydrolipoamide dehydrogenasemitochondrial, DLDH, E3, E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, EC 1.8.1, EC 1.8.1.4, GCSL, Glycine cleavage system L protein, glycine cleavage system protein L, lipoamide dehydrogenase, lipoamide reductase, lipoyl dehydrogenase
Gene Symbols DLD
Host Species Mouse
Immunogen Full length recombinant protein of human Dihydrolipoamide Dehydrogenase/DLD (NP_000099) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1738
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.