Learn More
Dihydrolipoamide Dehydrogenase/DLD Antibody (OTI6D5), Alexa Fluor™ 700, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Dihydrolipoamide Dehydrogenase/DLD |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | OTI6D5 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | 2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex), branched chain keto acid dehydrogenase complex, dihydrolipoamide dehydrogenase (E3 component of pyruvate dehydrogenase complex, dihydrolipoamide dehydrogenasemitochondrial, DLDH, E3, E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, EC 1.8.1, EC 1.8.1.4, GCSL, Glycine cleavage system L protein, glycine cleavage system protein L, lipoamide dehydrogenase, lipoamide reductase, lipoyl dehydrogenase |
| Gene Symbols | DLD |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.