Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopentyl ether, mixture of isomers, 96%

Catalog No. p-7081306
Click to view available options
Quantity:
25 mL
100 mL
500 mL

CAS: 544-01-4 | C10H22O | 158.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-01-4
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00008947
InChI Key AQZGPSLYZOOYQP-UHFFFAOYSA-N
Synonym diisopentyl ether, isoamyl ether, isopentyl ether, diisoamyl ether, isoamyl oxide, butane, 1,1'-oxybis 3-methyl, di-3-methylbutyl ether, di-iso-amyl ether, 1,1'-oxybis 3-methylbutane, 1,1'-oxylbis 3-methylbutane
PubChem CID 10989
IUPAC Name 3-methyl-1-(3-methylbutoxy)butane
SMILES CC(C)CCOCCC(C)C

Specifications

Density 0.78
Boiling Point 172°C to 174°C
Flash Point 46°C (115°F)
Refractive Index 1.407
Quantity 25 mL
UN Number UN3271
Beilstein 1698014
Merck Index 14,5118
Formula Weight 158.29
Percent Purity 96%
Chemical Name or Material Diisopentyl ether, Mixture of isomers
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 208-857-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.