Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl cyanomethylphosphonate, 97%

Catalog No. AAB2216306
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAB2216306

Supplier: Thermo Scientific Chemicals B2216306

Only null left
Add to Cart

CAS: 21658-95-7 | C8H16NO3P | 205.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21658-95-7
Molecular Formula C8H16NO3P
Molecular Weight (g/mol) 205.194
MDL Number MFCD00009812
InChI Key MQDIIJHQSUNBKA-UHFFFAOYSA-N
Synonym diisopropyl cyanomethyl phosphonate, diisopropyl cyanomethylphosphonate, diisopropyl cyanomethylphosphonate,, acmc-20al7r, diisopropoxyphosphinylacetonitrile, 2-di propan-2-yloxy phosphorylacetonitrile, diisopropyl cyanomethylphosphonate 5g, phosphonic acid,p-cyanomethyl-, bis 1-methylethyl ester
PubChem CID 88997
IUPAC Name 2-di(propan-2-yloxy)phosphorylacetonitrile
SMILES CC(C)OP(=O)(CC#N)OC(C)C

Specifications

Density 1.039
Boiling Point 108°C to 110°C (0.08 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4295
Quantity 5 g
UN Number UN2927
Beilstein 1946176
Formula Weight 205.19
Percent Purity 97%
Chemical Name or Material Diisopropyl cyanomethylphosphonate
Hazard Category H301+H311-H314-H330-H335
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 244-511-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.