Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethoxymethane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
Used to protect diols as their methylene acetals, using TMS-OTf/2,6-lutidine:
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used to protect diols as their methylene acetals, using TMS-OTf/2,6-lutidine:
Solubility
Soluble in water 20°C 330g/L.
Notes
Moisture Sensitive. Store away from oxidizing agents, moisture/water and acids. Store under inert gas.
Used to protect diols as their methylene acetals, using TMS-OTf/2,6-lutidine:
Solubility
Soluble in water 20°C 330g/L.
Notes
Moisture Sensitive. Store away from oxidizing agents, moisture/water and acids. Store under inert gas.
Specifications
Melting Point | -105°C |
Density | 0.863 |
Boiling Point | 41°C to 42°C |
Flash Point | −17°C (1°F) |
Refractive Index | 1.3545 |
Linear Formula | CH3OCH2OCH3 |
Quantity | 100 mL |
UN Number | UN1234 |
Beilstein | 1697025 |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H225 |
Hazard Statement | GHS H Statement H225-H319 Highly flammable liquid and vapor. Causes serious eye irritation. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYLAL |
EINECSNumber | 203-714-2 |
RTECSNumber | PA8750000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction