Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethyl-d6 sulfate, for NMR, 99+ atom % D
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
Specifications
Specifications
| Physical Form | Liquid |
| Color | Colorless |
| Refractive Index | 1.385 |
| Flash Point | 83°C |
| Quantity | 1 g |
| Packaging | Glass Bottle |
| Density | 1.3900g/mL |
| Deuterium (D) | 99.0% min. |
| Boiling Point | 60.0°C to 61.0°C (7.0mmHg) |
| Linear Formula | (CD3)2SO4 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Danger |
| missing translation for 'healthHazard2' | GHS H Statement: Fatal if inhaled. Suspected of causing genetic defects. Causes severe skin burns and eye damage. May cause an allergic skin reaction. Causes serious eye damage. Toxic if swallowed. May cause cancer. |
| missing translation for 'healthHazard3' | GHS P Statement: IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear eye protection/face protection. Use personal protective equipment as required. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction