Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl glutarate, 98%

Catalog No. p-7027533
Click to view available options
Quantity:
25 g
100 g

Catalog No. AAB2435914

Supplier: Thermo Scientific Chemicals B2435914

Only null left
Add to Cart

CAS: 1119-40-0 | C7H12O4 | 160.17 g/mol

Dimethyl glutarate is applied as paint and graffiti removers, nail polish removers, hand cleaners, adhesives, sealants and caulks, automotive cleaning products, bathroom and tile cleaners, general purpose cleaners, spot removers, floor maintenance products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl glutarate is applied as paint and graffiti removers, nail polish removers, hand cleaners, adhesives, sealants and caulks, automotive cleaning products, bathroom and tile cleaners, general purpose cleaners, spot removers, floor maintenance products.

Solubility
Soluble in water. (53 g/L)

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-40-0
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00008468
InChI Key XTDYIOOONNVFMA-UHFFFAOYSA-N
Synonym dimethyl glutarate, glutaric acid dimethyl ester, pentanedioic acid, dimethyl ester, methyl glutarate, glutaric acid, dimethyl ester, dbe 5, unii-1i9vfa346p, pentanedioic acid, 1,5-dimethyl ester, 1,5-dimethyl pentanedioate, pentanedioic acid dimethyl ester
PubChem CID 14242
SMILES COC(=O)CCCC(=O)OC

Specifications

Melting Point -37°C
Density 1.087
Boiling Point 213°C to 215°C
Flash Point 103°C (217°F)
Refractive Index 1.424
Quantity 25 g
Beilstein 1771444
Merck Index 14,4473
Solubility Information Soluble in water. (53g/L)
IUPAC Name dimethyl pentanedioate
Formula Weight 160.17
Percent Purity 98%
Chemical Name or Material Dimethyl glutarate
Show More Show Less
EINECSNumber 214-277-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.