Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Phthalate, 99%, (DMP), Reagents

Catalog No. 01337278
Click to view available options
:
Quantity: 500 mL

Catalog No. 01-337-278

Supplier: Reagents Holdings Llc C2196600500C

Only null left
Add to Cart
Add to Cart

Dimethyl Phthalate, 99%, (DMP), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 131-11-3
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
InChI Key NIQCNGHVCWTJSM-UHFFFAOYSA-N
Synonym Methyl phthalate, Dimethyl benzene-1,2-dicarboxylate
IUPAC Name 1,2-dimethyl benzene-1,2-dicarboxylate
SMILES COC(=O)C1=CC=CC=C1C(=O)OC

Specifications

Boiling Range 283°C
Melting Point -42°C
Color Colorless to Slight Yellow
Boiling Point 283°C
Melting Range -42°C
Packaging Glass Bottle
Quantity 500 mL
Specific Gravity 1.191
Grade Reagent
Physical Form Liquid
Chemical Name or Material Dimethyl Phthalate, 99%, (DMP)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.