Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfoxide-d{6}, 100% (Isotopic), contains 0.03% v/v TMS

Catalog No. p-7051939
Click to view available options
Quantity:
10 Ea.
2 Ea.

CAS: 2206-27-1 | C2H6OS | 84.17 g/mol

DMSO is a common polar solvent used in NMR, PCR, as a cryoprotectant, and as a universal solvent for many compounds. Deuterated DMSO is a common solvent used in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2206-27-1
Molecular Formula C2H6OS
Molecular Weight (g/mol) 84.17
MDL Number MFCD00002090
InChI Key IAZDPXIOMUYVGZ-WFGJKAKNSA-N
Synonym dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
PubChem CID 75151
IUPAC Name trideuterio(trideuteriomethylsulfinyl)methane
SMILES [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Packaging Prescored Ampule
Quantity 2 Ea.
Refractive Index 1.476
Density 1.18 g/mL
Flash Point 88°C (190°F)
Sensitivity Hygroscopic
Odor Characteristic
Melting Point 20°C
Boiling Point 190°C
Solubility Information Minimum isotopic purity 99.96%
Formula Weight 84.17
Percent Purity 100%
Concentration or Composition (by Analyte or Components) Contains 0.03% v/v TMS
Chemical Name or Material Dimethyl sulfoxide-d6, Isotopic
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 218-617-0
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.