Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethylarginine Dimethylaminohydrolase 1/DDAH1 Rabbit anti-Human, Rat, Clone: 1Q5F6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Dimethylarginine Dimethylaminohydrolase 1/DDAH1 |
---|---|
Clone | 1Q5F6 |
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Dimethylarginine Dimethylaminohydrolase 1/DDAH1 Monoclonal antibody specifically detects Dimethylarginine Dimethylaminohydrolase 1/DDAH1 in Human, Rat samples. It is validated for Western BlotSpecifications
Dimethylarginine Dimethylaminohydrolase 1/DDAH1 | |
Western Blot 1:500 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
DDAH-1, DDAHdimethylargininase-1, DDAHI, Dimethylargininase-1, dimethylarginine dimethylaminohydrolase 1FLJ25539, EC 3.5.3.18, FLJ21264, N(G), N(G)-dimethylarginine dimethylaminohydrolase 1, NG, NG-dimethylarginine dimethylaminohydrolase | |
A synthetic peptide corresponding to a sequence within amino acids 186-285 of human Dimethylarginine Dimethylaminohydrolase 1/DDAH1 (O94760). IAIGSSESAQKALKIMQQMSDHRYDKLTVPDDIAANCIYLNIPNKGHVLLHRTPEEYPESAKVYEKLKDHMLIPVSMSELEKVDGLLTCCSVLINKKVDS | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
1Q5F6 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
23576 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title