Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Dimethylenastron
Click to view available options
Quantity:
10 mg
50 mg
Description
Dimethylenastron is an inhibitor of mitotic motor kinesin Eg5 (IC50 = 200 nM). Arrests mitosis, triggers apoptosis and up-regulates Hsp-70 expression in multiple human myeloma cell lines. Also inhibits the growth of several types of cancer cell lines (EC50 values are 330, 603, 743 and 881 nM in HCT116, Htert-HME1, BxPC3, K562 and NCI-H1299 cancer cells, respectively). Suppresses the migration and invasion of PANC1 cells in vitro. Cytoprotective.
Specifications
Specifications
Chemical Name or Material | 2,3,4,6,7,8-Hexahydro-4-(3-hydroxyphenyl)-7,7-dimethyl-2-thioxo-5(1H)-quinazolinone |
CAS | 863774-58-7 |
Quantity | 10 mg |
Target | Kinesin Inhibitors |
Molecular Formula | C16H18N2O2S |
Formula Weight | 302.39 |
Purity | >99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction