Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfate, 99+%, AcroSeal™

Catalog No. p-4089403
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle

Catalog No. AC430831000

Supplier: Thermo Scientific Chemicals 430831000

Only null left
Add to Cart

CAS: 77-78-1 | C2H6O4S | 126.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-78-1
Molecular Formula C2H6O4S
Molecular Weight (g/mol) 126.13
MDL Number MFCD00008416
InChI Key VAYGXNSJCAHWJZ-UHFFFAOYSA-N
Synonym dimethyl sulphate, sulfuric acid, dimethyl ester, dimethyl monosulfate, dimethylsulfate, dimethylsulfat, sulfate dimethylique, sulfuric acid dimethyl ester, dimethylsulfaat, dimetilsolfato, dms methyl sulfate
PubChem CID 6497
ChEBI CHEBI:59050
IUPAC Name dimethyl sulfate
SMILES COS(=O)(=O)OC

Specifications

Melting Point -32.0°C
Density 1.3300g/mL
Boiling Point 188.0°C
Flash Point 83°C
Assay Percent Range 99+%
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.3860 to 1.3880
Linear Formula (CH3)2SO4
Quantity 100 mL
Merck Index 15,328
Specific Gravity 1.33
Solubility Information Solubility in water: 2.8g/100mL (18°C). Other solubilities: hydrolysis rapidly at 18°C or above in water,soluble in ehter,dioxane,acetone and aromatic,hydrocarbons,sparingly soluble in carbon disulfide and,aliphatic hydrocarbons
Viscosity 1.81 mPa.s (20°C)
Formula Weight 126.13
Percent Purity 99+%
Chemical Name or Material Dimethyl sulfate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
Suspected of causing genetic defects.
Fatal if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause respiratory i
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 201-058-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.