Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIMT1L Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB047261 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB047261 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB047261 Supplier Novus Biologicals Supplier No. NBP297740F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DIMT1L Polyclonal specifically detects DIMT1L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DIMT1L
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase, DIM1, DIM1 dimethyladenosine transferase 1-like, DIM1 dimethyladenosine transferase 1-like (S. cerevisiae), DIMT1dimethyladenosine transferase, EC 2.1.1, EC 2.1.1.14, EC 2.1.1.183,18S rRNA dimethylase, HSA9761, HUSSY5, N'-adenosyl(rRNA) dimethyltransferase, probable dimethyladenosine transferase, S-adenosylmethionine-6-N'
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DIMT1L fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 27292
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.