Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIO3 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP105767R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105767R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105767R Supplier Novus Biologicals Supplier No. NBP105767R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DIO3 Polyclonal specifically detects DIO3 in Human, Mouse, Rat, Hamster, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen DIO3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 5DIII, D3, deiodinase, iodothyronine, type III, DIOIII, EC 1.97.1, EC 1.97.1.11, ITDI3, placental type, thyroxine deiodinase type III (selenoprotein), Type 3 DI, type 3 iodothyronine selenodeiodinase, type III iodothyronine deiodinase, type-III 5' deiodinase, Type-III 5'-deiodinase
Gene Symbols DIO3
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of rat DIO3 (within residues 250-300). [Swiss-Prot# P49897]
Molecular Weight of Antigen 31 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Growth and Development, Lipid and Metabolism, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1735
Test Specificity Human, rat, hamster and mouse. Rabbit reactivity via customer review.
Target Species Human, Mouse, Rat, Hamster, Rabbit
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.