Learn More
Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns

Description
Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.
- Achieve n, n-1 resolution for oligonucleotides
- Resolve oligonucleotides with secondary structures
- Assay phosphorothioate purity
- Selectivity control with eluent pH, salt, and solvent
- Resolve RNA with aberrant (2' – 5') links from normal SSRNA
Specifications
Specifications
| Column Format | Guard Column |
| Diameter (Metric) | 2 mm |
| For Use With | HPAE-PAD |
| Length (Metric) | 50 mm |
| Max. Pressure | 4000 psi (275 bar) |
| Particle Size | 8 μm |
| pH | 2.5 to 12.5 |
| Pore Size | Nonporous |
| Quantity | Each |
| Temperature | 85°C |
| Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.