Learn More
Eisco™ Diorite Specimen
SCI_ED
Catalog No. S24750 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24750 1 Ea. 2 to 3 cm
S26632 10/Pk. 2 to 3 cm
S24755 12/Pk. 2 to 3 cm
S26670 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24750 Supplier Eisco™ Supplier No. ESNG0042
Add to Cart
Add to Cart
Diorite Specimen straddles the line between mafic and felsic rock chemistry.
  • Presence of both mafic and felsic minerals make diorite example of intermediate igneous chemistry
  • For use in lessons in bulk chemistry
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Large crystal size accommodates mineral identification with hand lens (not included)
  • The minerals within diorite can be readily identified due to their large size
Specifications
Product Type Igneous Rock
For Use With (Application) Lessons in bulk chemistry
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Diorite