Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIP13B Antibody, Novus Biologicals™
SDP

Catalog No. NBP216165 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216165 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216165 Supplier Novus Biologicals Supplier No. NBP216165
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

DIP13B Polyclonal specifically detects DIP13B in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen DIP13B
Applications Western Blot
Classification Polyclonal
Concentration 0.6mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q8NEU8
Gene Alias Adapter protein containing PH domain, PTB domain and leucine zipper motif 2, adaptor protein containing PH domain, PTB domain and leucine zipper motif 2, adaptor protein, phosphotyrosine interaction, PH domain and leucine zippercontaining 2, DCC-interacting protein 13-beta, DIP13BDIP13 beta, Dip13-beta, FLJ10659dip13-beta
Gene Symbols APPL2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human DIP13B. The exact sequence is proprietary.
Molecular Weight of Antigen 74 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55198
Test Specificity Human. Expected to cross react based on sequence identity: Mouse (94%), Rat (93%), Xenopus laevis (87%), Bovine (95%), Xenopus tropicalis (87%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.