Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DIP2A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
Antigen | DIP2A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15690920
![]() |
Novus Biologicals
NBP15690920UL |
20 μL |
Each for $158.00
|
|
|||||
NBP156909
![]() |
Novus Biologicals
NBP156909 |
100 μL |
Each for $487.50
|
|
|||||
Description
DIP2A Polyclonal specifically detects DIP2A in Human samples. It is validated for Western Blot.Specifications
DIP2A | |
Polyclonal | |
Rabbit | |
Q14689 | |
23181 | |
Synthetic peptides corresponding to DIP2A(DIP2 disco-interacting protein 2 homolog A (Drosophila)) The peptide sequence was selected from the N terminal of DIP2A. Peptide sequence PLKEFFVDDFEELLEVQQPDPNQPKPEGSETSVLRGEPLTAGVPRPPSLL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Dip2, DIP2 disco-interacting protein 2 homolog A (Drosophila), DIP2 homolog A, DIP2C21orf106, disco-interacting protein 2 homolog A, disco-interacting protein 2A, KIAA0184chromosome 21 open reading frame 106 | |
DIP2A | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title