Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl Chlorophosphate 95.0+%, TCI America™
SDP

Catalog No. D1059100G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
100 g
500 g

Catalog No. D1059100G

Supplier: TCI America D1059100G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2524-64-3
Molecular Formula C12H10ClO3P
Molecular Weight (g/mol) 268.633
MDL Number MFCD00003030
InChI Key BHIIGRBMZRSDRI-UHFFFAOYSA-N
Synonym diphenyl chlorophosphate, diphenyl phosphorochloridate, diphenylchlorophosphate, phosphorochloridic acid, diphenyl ester, chlorodiphenyl phosphate, o,o-diphenyl chlorophosphate, chlorodiphenoxyphosphine oxide, diphenoxychlorophosphine oxide, diphenyl chlorophosphonate, diphenylphosphoric acid monochloride
PubChem CID 75654
IUPAC Name [chloro(phenoxy)phosphoryl]oxybenzene
SMILES C1=CC=C(C=C1)OP(=O)(OC2=CC=CC=C2)Cl

Specifications

Color Yellow
Boiling Point 316°C
Quantity 100 g
UN Number 3265
Formula Weight 268.63
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diphenyl Chlorophosphate
EINECSNumber (3)-3358
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.