Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl diselenide, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1666-13-3
C12H10Se2
312.13
MFCD00003001
YWWZCHLUQSHMCL-UHFFFAOYSA-N
diphenyl diselenide,diselenide, diphenyl,phenyl diselenide,1,2-diphenyldiselane,diphenyldiselenide,bis phenylselenide,phenyldiselanyl benzene,diphenyldiselane,ccris 9227,acmc-209duq
15460
(phenyldiselanyl)benzene
C1=CC=C(C=C1)[Se][Se]C2=CC=CC=C2
1666-13-3
C12H10Se2
312.13
MFCD00003001
YWWZCHLUQSHMCL-UHFFFAOYSA-N
diphenyl diselenide,diselenide, diphenyl,phenyl diselenide,1,2-diphenyldiselane,diphenyldiselenide,bis phenylselenide,phenyldiselanyl benzene,diphenyldiselane,ccris 9227,acmc-209duq
15460
(phenyldiselanyl)benzene
C1=CC=C(C=C1)[Se][Se]C2=CC=CC=C2
Melting Point 60.0°C to 63.0°C
Color Yellow
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Assay Percent Range 98.5% min. (GC)
Linear Formula (C6H5)2Se2
Beilstein 06, 346
Fieser 05,272; 06,235; 07,136; 09,199; 10,170; 11,219; 13,125; 16,152; 17,352
Solubility Information Solubility in water: insoluble in water.
Formula Weight 312.13
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Diphenyl diselenide
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and

GHS Signal Word: Danger

EINECSNumber : 216-780-2