Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl sulfone, 99+%

Catalog No. p-7044788
Click to view available options
Quantity:
250 g
1000 g

CAS: 127-63-9 | C12H10O2S | 218.27 g/mol

It is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-63-9
Molecular Formula C12H10O2S
Molecular Weight (g/mol) 218.27
MDL Number MFCD00007548
InChI Key KZTYYGOKRVBIMI-UHFFFAOYSA-N
Synonym diphenyl sulfone, phenyl sulfone, sulfonyldibenzene, diphenylsulfone, sulfobenzide, diphenyl sulphone, benzene, 1,1'-sulfonylbis, sulfone, diphenyl, 1,1'-sulfonyldibenzene, phenyl sulphone
PubChem CID 31386
ChEBI CHEBI:78360
IUPAC Name benzenesulfonylbenzene
SMILES O=S(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 123°C to 130°C
Density 1.36
Boiling Point 377°C to 379°C
Flash Point 184°C (363°F)
Odor Odorless
Quantity 250 g
Beilstein 1910573
Merck Index 14,3238
Solubility Information Insoluble in water.
Formula Weight 218.27
Percent Purity ≥99%
Chemical Name or Material Diphenyl sulfone
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 204-853-1
RTECSNumber SX2400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.