Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylcarbazide, 1% (w/v) Alcoholic, Ricca Chemical
SDP

Catalog No. 259116 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
259116 500 mL Amber Glass Bottle
25914 120 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 259116 Supplier Ricca Chemical Company Supplier No. 259116
Add to Cart
Add to Cart

Chemical Identifiers

CAS 140-22-7 , 64-17-5 , 67-56-1
Molecular Formula C13H14N4O
Molecular Weight (g/mol) 242.28
InChI Key KSPIHGBHKVISFI-UHFFFAOYSA-N
Synonym 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
PubChem CID 8789
ChEBI CHEBI:4641
IUPAC Name 1,3-bis(phenylamino)urea
SMILES O=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1

Specifications

Color Yellow
CAS Min % 1.21, 97.0
CAS Max % 1.26, 100.0
Percent Purity 1%
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Alcoholic
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 1% (w/v)
Chemical Name or Material Diphenylcarbazide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.