Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphtheria Toxin Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10062518 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062518 0.5 mL
NB017488 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10062518 Supplier Novus Biologicals Supplier No. NB10062518
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Diphtheria Toxin Polyclonal specifically detects Diphtheria Toxin in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Diphtheria Toxin
Applications ELISA
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation PBS with 0.1% Sodium Azide
Gene Alias Diphtheria Toxin, tox
Host Species Goat
Immunogen Purified diphtheria toxoid
Purification Method Affinity Purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product reacts with the diphtheria toxin/toxoid. Diphtheria toxin is an exotoxin secreted by various strains of Corynebacterium diphtheriae that is the cause of Diphtheria. The active toxin is a single chain of 535 amino acids composed of 3 distinct structural/functional domains; and N-terminal ADP-ribosyltransferase (catalytic domain); a region which facilitates the delivery of the catalytic domain across the cell membrane (transmembrane domain) and the eukaryotic cell receptor binding domain.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.