Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphtheria Toxoid Antibody (17.6B9) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP22359601 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP22359601 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP22359601 Supplier Novus Biologicals Supplier No. NBP2235960.1ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Diphtheria Toxoid Monoclonal specifically detects Diphtheria Toxoid in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Diphtheria Toxoid
Applications ELISA
Classification Monoclonal
Clone 17.6B9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide
Gene Alias C. Diphtheria toxin, Corynebacterium diphtheriae toxin, Corynebacterium diphtheriae toxoid, DIP0222, Diptheria toxin, Diptheriae toxin, DTx
Host Species Mouse
Immunogen Diphtheria toxoid prepared from toxin purified from C. diphtheriae subsp. intermedius Park-Williams no. 8 by formaldehyde inactivation (2), and adsorbed to aluminum hydroxide gel.
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity It reacts with diphtheria toxoid and also neutralizes diphtheria toxin as determined in a VERO cell assay
Target Species Bacteria
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.