Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DIPPA hydrochloride
GREENER_CHOICE

Catalog No. p-4783420
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective and irreversible κ antagonist

An irreversible and selective antagonist at the κ receptor, with persistent effect in vivo.

Chemical Identifiers

CAS 155512-52-0
Molecular Formula C22H24Cl3N3OS

Specifications

Quantity 10 mg
Formula Weight 484.87
Purity >99%
Molecular Weight (g/mol) 484.864
Chemical Name or Material DIPPA hydrochloride
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.